missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUSAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Especificaciones
Antígeno | NUSAP1 |
---|---|
Concentración | 0.18 mg/mL |
Aplicaciones | Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot |
Clasificación | Polyclonal |
Conjugado | Unconjugated |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16819493
|
Proteintech
12024-1-AP-20UL |
20 μL |
140.00€
20 microlitros |
Fecha estimada de envío: 21-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16809493
|
Proteintech
12024-1-AP-150UL |
150 μL |
399.00€
150 microlitros |
Fecha estimada de envío: 21-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Ubiquitin: exists either covalently attached to another protein, or free (uchored). When covalently bound, it is conjugated to target proteins via an isopeptide bond either as a monomer (monoubiquitin), a polymer linked via different Lys residues of the ubiquitin (polyubiquitin chains) or a linear polymer linked via the initiator Met of the ubiquitin (linear polyubiquitin chains). Polyubiquitin chains, when attached to a target protein, have different functions depending on the Lys residue of the ubiquitin that is linked: Lys-6-linked may be involved in DNA repair; Lys-11-linked is involved in ERAD (endoplasmic reticulum-associated degradation) and in cell-cycle regulation; Lys-29-linked is involved in lysosomal degradation; Lys-33-linked is involved in kinase modification; Lys-48-linked is involved in protein degradation via the proteasome; Lys-63-linked is involved in endocytosis, DNA-damage responses as well as in signaling processes leading to activation of the transcription factor NF-kappa-B. Linear polymer chains formed via attachment by the initiator Met lead to cell signaling. Ubiquitin is usually conjugated to Lys residues of target proteins, however, in rare cases, conjugation to Cys or Ser residues has been observed. When polyubiquitin is free (uchored-polyubiquitin), it also has distinct roles, such as in activation of protein kinases, and in signaling. 60S ribosomal protein L40: component of the 60S subunit of the ribosome. Ribosomal protein L40 is essential for translation of a subset of cellular transcripts, and especially for cap-dependent translation of vesicular stomatitis virus mRNAs.Especificaciones
NUSAP1 | |
Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q9BXS6, Q9ERH4 | |
108907, 311336, 51203 | |
NUSAP1 Fusion Protein Ag2654 | |
Primary | |
-20°C | |
NUSAP1 |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ANKT, BM037, LNP, NuSAP, NUSAP1, PRO0310, PRO0310p1, Q0310, SAPL | |
NUSAP1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |