Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates, HydroCell
These microplates are optimized for robotics, automated readers and liquid handling systems. Ideal for antibiotic screens, cell-based assays and screening compounds.
Marca: Thermo Scientific™ 174907
Detalles adicionales : Peso : 4.43000kg
Descripción
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
Especificaciones
Todas las microplacas fabricadas por Nunc cumplen las recomendaciones de American National Standards (ANSI) (ANSI/SBS 1-2004) | |
Sí | |
8 x 12 | |
Nunc | |
Microplate | |
96 | |
HydroCell | |
8 | |
8/Pk. |
Standard | |
Placa de 96 pocillos, HydroCell, transparente, con tapa, estéril (paquete de 1, caja de 8) | |
8 Pk., 8 Cases | |
Transparente | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
8 |
Exención de responsabilidad de la restricción de uso para investigación interna: Cualquier restricción impuesta al comprador para utilizar el producto o productos únicamente para fines de investigación interna no se aplica al producto o productos que aparecen en las páginas web asociadas a esta declaración.