missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ 1536-Well, Cell Culture-Treated, Flat-Bottom Microplate

Color:White; Sterility:Sterile; Lid:With Lid

Marca:  Thermo Scientific™ 164709

Detalles adicionales : Peso : 5.44300kg

Código de producto. 10608344

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Work with small volumes and screen 4X as many samples in one plate. Thermo Scientific™ Nunc 1536 Well High Base Microplates are ideal for high-throughput screening; the skirted design around the base of the plate is optimal for robotic handling.

  • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth.
  • Polystyrene
  • Working range: 2–10μL/well
  • Customized bar coding is available
Especificaciones

Especificaciones

All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
With Lid
25 Pk., 100 Cases
White
Skirted
13.4 μL
10 mL
Cell Culture
Flat Bottom (F-well)
25
Standard
32 x 48
Nunc
Microplate
Polystyrene
1536
Sterile
13.4 μL
100
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales