Learn More
NUDT17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA556365
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: RLSWGLPKYH HIVLYLLVIS QESQQQLQAR IQPNPNEVSA LMWLTPDVAA AVAAAEDGTE TPGLLPQDLP PSV Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.
Probably mediates the hydrolysis of some nucleoside diphosphate derivatives.Especificaciones
NUDT17 | |
Polyclonal | |
Unconjugated | |
NUDT17 | |
2410015C20Rik; nucleoside diphosphate-linked moiety X motif 17; nudix (nucleoside diphosphate linked moiety X)-type motif 17; nudix hydrolase 17; nudix motif 17; NUDT17; RGD1565469 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
200035 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P0C025 | |
NUDT17 | |
Recombinant protein corresponding to Human NUDT17 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only