missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17487-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Especificaciones
NUDT12 | |
Polyclonal | |
Unconjugated | |
NUDT12 | |
Nudix motif 12, NUDT12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
367323, 67993, 83594 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BQG2, Q9DCN1 | |
Nudt12 | |
NUDT12 Fusion Protein Ag11557 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |