Learn More
NUDC Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10681-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
NudC was first identified as a regulator of nuclear movement in the asexual reproductive cycle of the filamentous fungus Aspergillus nidulans. Human NUDC is a nuclear movement protein that associates with dynein.Especificaciones
NUDC | |
Polyclonal | |
Unconjugated | |
NUDC | |
HNUDC, MNUDC, Nuclear migration protein nudC, NUDC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10726 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y266 | |
NUDC | |
NUDC Fusion Protein Ag0998 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |