Learn More
NUBP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA559684
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: STISTELALA LRHAGKKVGI LDVDLCGPSI PRMLGAQGRA VHQCDRGWAP VF Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
NUBP2 Polyclonal antibody specifically detects NUBP2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
NUBP2 | |
Polyclonal | |
Unconjugated | |
NUBP2 | |
CFD1; Cytosolic Fe-S cluster assembly factor NUBP2; cytosolic Fe-S cluster assembly factor NUBP2 {ECO:0000255; D17Wsu11e; HAMAP-Rule:MF_03039}; homolog of yeast cytosolic Fe-S cluster deficient 1; NBP 2; NBP 2 {ECO:0000255; NUBP1; Nubp2; nucleotide binding protein 2; nucleotide binding protein 2 (MinD homolog, E. coli); Nucleotide-binding protein 2; nucleotide-binding protein 2 {ECO:0000255 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10101 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y5Y2 | |
NUBP2 | |
Recombinant protein corresponding to Human NUBP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only