missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-350 of E.coli MYH The Recombinant E. coli MYH Protein is derived from E. coli. The Recombinant E. coli MYH Protein has been validated for the following applications: SDS-Page.
Especificaciones
Especificaciones
| Concentración | 0.25 mg/mL |
| Para utilizar con (aplicación) | SDS-PAGE |
| Formulación | 20 mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 20% glycerol |
| ID de gen (Entrez) | 4595 |
| Peso molecular | M.W. (theoretical): 41.5 kDa |
| Nombre | MYH Protein |
| Método de purificación | SDS-PAGE |
| Cantidad | 0,01 mg |
| Fuente | E. Coli |
| Inmunógeno | mutY, 1-350aa, E.coli, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMQASQFS AQVLDWYDKY GRKTLPWQID KTPYKVWLSE VMLQQTQVAT VIPYFERFMA RFPTVTDLAN APLDEVLHLW TGLGYYARAR NLHKAAQQVA TLHGGKFPET FEEVAALPGV GRSTAGAILS LSLGKHFPIL DGNVKRVLAR CYAVSGWPGK KEVENKLWSL SEQVTPAVGV ERFNQAMMDL GAMICTRSKP KCSLCPLQNG CIAAANNSWA LYPGKKPKQT LPERTGYFLL LQHEDEVLLA QRPPSGLWGG LYCFPQFADE ESLRQWLAQR QIAADNLTQL TAFRHTFSHF HLDIVPMWLP VSSFTGCMDE GNALWYNLAQ PPSVGLAAPV ERLLQQLRTG APV |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?