Learn More
Novus Biologicals™ Recombinant Human ERp57/PDIA3 His Protein
Código de producto. 18781014 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0,1 mg
Tamaño de la unidad:
0.10 miligramo
18781014 0,1 mg 0.10 miligramo
1 options
Código de producto. 18781014
Marca: Novus Biologicals™ NBP137081
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 25-505 of Human ERp57/PDIA3 The Recombinant Human ERp57/PDIA3 Protein is derived from E. coli. The Recombinant Human ERp57/PDIA3 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Especificaciones
Cantidad 0,1 mg