Learn More
Novus Biologicals™ Fascin Protein
Código de producto. 18747553 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0,1 mg
0,5 mg
Tamaño de la unidad:
0.10 miligramo
0.50 miligramo
18747553 0,1 mg 0.10 miligramo
18799213 0,5 mg 0.50 miligramo
2 options
Código de producto. 18747553
Marca: Novus Biologicals™ NBP1453130.1mg
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-493 of Human Fascin The Recombinant Human Fascin Protein is derived from E. coli. The Recombinant Human Fascin Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Especificaciones
Cantidad 0,1 mg