Learn More
Novus Biologicals™ ACBP Protein
Código de producto. 18766213 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0,1 mg
0,5 mg
Tamaño de la unidad:
0.10 miligramo
0.50 miligramo
18766213 0,5 mg 0.50 miligramo
18222922 0,1 mg 0.10 miligramo
2 options
Código de producto. 18766213
Marca: Novus Biologicals™ NBP1508330.5mg
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-87 of Human ACBP The Recombinant Human ACBP Protein is derived from E. coli. The Recombinant Human ACBP Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Cantidad 0,5 mg