missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK1.1 (CD161) Mouse, anti-Mouse, PE, Clone: PK136, STEMCELL Technologies
Mouse Monoclonal Antibody
en la oferta
Especificaciones
| Antígeno | NK1.1 (CD161) |
|---|---|
| Clon | PK136 |
| Aplicaciones | CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay |
| Clasificación | Monoclonal |
| Conjugado | PE |
Descripción
- The PK136 antibody reacts with murine NK1.1 (CD161), an ∼80 kDa homodimeric type 2 transmembrane glycoprotein and C-type lectin receptor expressed on NK cells, NK-T cells, and rare subsets of T cells and monocytes in select strains of mice
- NK1.1 is encoded by the Klrb1b and Klrb1c genes, which specify CD161b and CD161c polypeptides, respectively
- CD161b is expressed only by Swiss NIH and SJL mice, and CD161c by certain strains such as C57BL, FVB/N, and NZB (but not A, AKR, BALB/c, CBA/J, C3H, C57BR, C58, DBA/1, DBA/2, NOD, SJL, or 129)
- NK1.1 has functional roles in modulating several processes, including the activation and proliferation of NK cells, induction of interferon-γ production, and release of cytotoxic granules
- Its expression on the cell surface is specifically upregulated by IL-12
- For detection of NK cells in NK1.1- strains, Anti-Mouse CD49b Antibody, Clone DX5 (Catalog #60020) is recommended
- DX5 recognizes CD49b (integrin α2), another commonly used NK cell marker
Especificaciones
| NK1.1 (CD161) | |
| CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| PE | |
| Mouse |
| PK136 | |
| Monoclonal | |
| Mouse | |
| Primary |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto