missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK1.1 (CD161) Mouse, anti-Mouse, PE, Clone: PK136, STEMCELL Technologies
Descripción
- The PK136 antibody reacts with murine NK1.1 (CD161), an ∼80 kDa homodimeric type 2 transmembrane glycoprotein and C-type lectin receptor expressed on NK cells, NK-T cells, and rare subsets of T cells and monocytes in select strains of mice
- NK1.1 is encoded by the Klrb1b and Klrb1c genes, which specify CD161b and CD161c polypeptides, respectively
- CD161b is expressed only by Swiss NIH and SJL mice, and CD161c by certain strains such as C57BL, FVB/N, and NZB (but not A, AKR, BALB/c, CBA/J, C3H, C57BR, C58, DBA/1, DBA/2, NOD, SJL, or 129)
- NK1.1 has functional roles in modulating several processes, including the activation and proliferation of NK cells, induction of interferon-γ production, and release of cytotoxic granules
- Its expression on the cell surface is specifically upregulated by IL-12
- For detection of NK cells in NK1.1- strains, Anti-Mouse CD49b Antibody, Clone DX5 (Catalog #60020) is recommended
- DX5 recognizes CD49b (integrin α2), another commonly used NK cell marker
Especificaciones
Especificaciones
| Antígeno | NK1.1 (CD161) |
| Aplicaciones | CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay |
| Clasificación | Monoclonal |
| Clon | PK136 |
| Conjugado | PE |
| Especie del huésped | Mouse |
| Cantidad | 100 μg |
| Primario o secundario | Primary |
| Especies diana | Mouse |
| Tipo de célula | NK Cells |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?