missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) Sulfate Hexahydrate, Reagent Grade, Honeywell™
Reagent Grade, powder or crystals
77.69€ - 174.20€
Identificadores de productos químicos
CAS | 10101-97-0 |
---|---|
Fórmula molecular | H12NiO10S |
Peso molecular (g/mol) | 262.839 |
Número MDL | MFCD00149813 |
Clave InChI | RRIWRJBSCGCBID-UHFFFAOYSA-L |
Sinónimo | nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 |
PubChem CID | 5284429 |
ChEBI | CHEBI:53437 |
Nombre IUPAC | níquel(2+); sulfato; hexahidrato |
SMILES | O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Descripción
- ≥95% for general laboratory use.
Identificadores de productos químicos
10101-97-0 | |
262.839 | |
RRIWRJBSCGCBID-UHFFFAOYSA-L | |
5284429 | |
níquel(2+); sulfato; hexahidrato |
H12NiO10S | |
MFCD00149813 | |
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
CHEBI:53437 | |
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Especificaciones
Nickel(II) sulfate hexahydrate | |
10101-97-0 | |
4 to 6 (at 20°C) | |
H12NiO10S | |
MFCD00149813 | |
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] | |
262.839 | |
CHEBI:53437 | |
Odorless |
53°C | |
Verde | |
Crystalline | |
NiSO4 · 6H2O | |
UN3077 | |
RRIWRJBSCGCBID-UHFFFAOYSA-L | |
níquel(2+); sulfato; hexahidrato | |
5284429 | |
262.85g/mol | |
1.95g/cu.cm at 20°C; Vapor Density: 2.07 (vs air) |
Seguridad y manipulación
P201-P261-P273-P280-P284-P304 + P340 + P312
missing translation for 'dotInformation' : 9/PGIII
missing translation for 'rtecsNumber' : 232-104-9