missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGFRAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10446-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
May be a signaling adapter molecule involved in p75NTR-mediated apoptosis induced by NGF. Plays a role in zinc-triggered neuronal death. May play an important role in the pathogenesis of neurogenetic diseases.Especificaciones
NGFRAP1 | |
Polyclonal | |
Unconjugated | |
BEX3 | |
Bex, BEX3, DXS6984E, HGR74, NADE, NGFRAP1, Protein BEX3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117089, 12070, 27018 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q00994, Q6PDU5, Q9WTZ9 | |
Bex3 | |
NGFRAP1 Fusion Protein Ag0378 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |