Learn More
NDUFC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 23842-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.Especificaciones
NDUFC1 | |
Polyclonal | |
Unconjugated | |
NDUFC1 | |
CI KFYI, Complex I KFYI, KFYI, NDUFC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4717 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43677 | |
NDUFC1 | |
NDUFC1 Fusion Protein Ag20835 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |