missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDST1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26203-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Essential bifunctional enzyme that catalyzes both the N- deacetylation and the N-sulfation of glucosamine (GlcNAc) of the glycosaminoglycan in heparan sulfate. Modifies the GlcNAc-GlcA disaccharide repeating sugar backbone to make N-sulfated heparosan, a prerequisite substrate for later modifications in heparin biosynthesis. Plays a role in determining the extent and pattern of sulfation of heparan sulfate. Compared to other NDST enzymes, its presence is absolutely required. Participates in biosynthesis of heparan sulfate that can ultimately serve as L- selectin ligands, thereby playing a role in inflammatory response.Especificaciones
NDST1 | |
Polyclonal | |
Unconjugated | |
NDST1 | |
HSST, HSST1, N HSST 1, NDST 1, NDST1, NST1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15531, 3340 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P52848, Q3UHN9 | |
NDST1 | |
NDST1 Fusion Protein Ag24422 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |