Learn More
N,N-Dimetilhexanamida, 98 %, Thermo Scientific Chemicals
Catalog Number 11400408
Click to view available options
Cantidad:
5 g
25 g
100 g
5830-30-8
C8H17NO
143.23
MFCD00043671
OAERLTPBKQBWHJ-UHFFFAOYSA-N
hexanamide, n,n-dimethyl, n,n-dimethylcaproamide, n,n-dimethylhexanoamide, nn-dimethylhexanamide, nn-dimethylcaproamide, n,n-dirnethylhexanamide, n,n-dimethyl-hexanamide, hexanamide,n,n-dimethyl, acmc-1atq3, n,n-dimethylhexanamide, tech
22084
N,N-Dimetilhexanamida
CCCCCC(=O)N(C)C
5830-30-8
C8H17NO
143.23
MFCD00043671
OAERLTPBKQBWHJ-UHFFFAOYSA-N
hexanamide, n,n-dimethyl, n,n-dimethylcaproamide, n,n-dimethylhexanoamide, nn-dimethylhexanamide, nn-dimethylcaproamide, n,n-dirnethylhexanamide, n,n-dimethyl-hexanamide, hexanamide,n,n-dimethyl, acmc-1atq3, n,n-dimethylhexanamide, tech
22084
N,N-Dimetilhexanamida
CCCCCC(=O)N(C)C
N,N-Dimethylhexanamide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
La N,N-dimetilhexanamida se utiliza como producto intermedio farmacéutico.
Solubilidad
Inmiscible o difícil de mezclar.
Notas
Mantener el recipiente bien sellado. Almacénese en un lugar fresco y seco en recipientes herméticos. Incompatible con agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N,N-Dimethylhexanamide
Densidad 0.89
Punto de ebullición 80°C (1 mmHg)
Índice de refracción 1.449
Cantidad 25 g
Beilstein 1747074
Información de solubilidad Not miscible or difficult to mix.
Formula Weight (peso de la fórmula) 143.23
Porcentaje de pureza 98%