missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-N-metil-L-alanina, 95 %, Thermo Scientific Chemicals
86.50€ - 1009.00€
Identificadores de productos químicos
CAS | 84000-07-7 |
---|---|
Fórmula molecular | C19H19NO4 |
Peso molecular (g/mol) | 325.36 |
Número MDL | MFCD00153384 |
Clave InChI | JOFHWKQIQLPZTC-UHFFFAOYNA-N |
Sinónimo | fmoc-n-me-ala-oh, fmoc-n-methyl-l-alanine, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino propanoic acid, fmoc-nalpha-methyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino propanoic acid, l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino propanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-alanine, fmoc-l-meala-oh, ambotzfaa1395 |
PubChem CID | 688634 |
Nombre IUPAC | Ácido (2S)-2-[9H-fluor-9-ilmetoxicarbonil(metil)amino]propanoico |
SMILES | CC(N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15470089
|
Thermo Scientific Alfa Aesar
H63411.06 |
5 g |
86.50€
5 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
15480089
|
Thermo Scientific Alfa Aesar
H63411.14 |
25 g |
294.00€
25 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
15490089
|
Thermo Scientific Alfa Aesar
H63411.22 |
100 g |
1009.00€
100 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Also used in N-methyl amino acids for peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Also used in N-methyl amino acids for peptide synthesis.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Identificadores de productos químicos
84000-07-7 | |
325.36 | |
JOFHWKQIQLPZTC-UHFFFAOYNA-N | |
688634 | |
CC(N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
C19H19NO4 | |
MFCD00153384 | |
fmoc-n-me-ala-oh, fmoc-n-methyl-l-alanine, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino propanoic acid, fmoc-nalpha-methyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino propanoic acid, l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino propanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-alanine, fmoc-l-meala-oh, ambotzfaa1395 | |
Ácido (2S)-2-[9H-fluor-9-ilmetoxicarbonil(metil)amino]propanoico |
Especificaciones
N-Fmoc-N-methyl-L-alanine | |
84000-07-7 | |
MFCD00153384 | |
fmoc-n-me-ala-oh, fmoc-n-methyl-l-alanine, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino propanoic acid, fmoc-nalpha-methyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino propanoic acid, l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino propanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-alanine, fmoc-l-meala-oh, ambotzfaa1395 | |
JOFHWKQIQLPZTC-UHFFFAOYNA-N | |
Ácido (2S)-2-[9H-fluor-9-ilmetoxicarbonil(metil)amino]propanoico | |
688634 | |
95% |
140°C | |
C19H19NO4 | |
4329998 | |
Slightly soluble in water. | |
CC(N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
325.36 | |
325.36 | |
Sólido |
Seguridad y manipulación
missing translation for 'tsca' : No
missing translation for 'storageNote' : De 2 °C a 8 °C
Recommended Storage : Mantener frío
RUO – Research Use Only