missing translation for 'onlineSavingsMsg'
Learn More

N-benciloxicarbonil-L-norleucina, 98+%, Thermo Scientific™

Catalog Number 11322856
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
39608-30-5
C14H19NO4
265.31
MFCD00063170
NMYWMOZOCYAHNC-UHFFFAOYNA-N
z-nle-oh, z-l-norleucine, 2s-2-benzyloxy carbonyl amino hexanoic acid, cbz-l-norleucine, z-norleu-oh, ambotzzaa1231, z-l-nle, z-l-2-aminohexanoic acid, benzyloxycarbonyl-l-norleucine, n-benzyloxycarbonyl-l-norleucine
6993403
ácido (2S)2-(fenilmetoxicarbonilamino)hexanoico
CCCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Este artículo no se puede devolver. Vea la política de devoluciones
39608-30-5
C14H19NO4
265.31
MFCD00063170
NMYWMOZOCYAHNC-UHFFFAOYNA-N
z-nle-oh, z-l-norleucine, 2s-2-benzyloxy carbonyl amino hexanoic acid, cbz-l-norleucine, z-norleu-oh, ambotzzaa1231, z-l-nle, z-l-2-aminohexanoic acid, benzyloxycarbonyl-l-norleucine, n-benzyloxycarbonyl-l-norleucine
6993403
ácido (2S)2-(fenilmetoxicarbonilamino)hexanoico
CCCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-Benzyloxycarbonyl-L-norleucine
Punto de fusión 56°C to 58°C
Cantidad 250 mg
Beilstein 3210973
Rotación óptica −10° (c=1 in Methanol)
Formula Weight (peso de la fórmula) 265.3
Porcentaje de pureza ≥98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'