Learn More
Gilson™ MyPIPETMAN™ Enterprise Multichannel Pipettes
The Enterprise pipettes combine reliability, ergonomics, and personalization options to ensure efficiency and answer your ranging pipetting needs.
en la oferta
Especificaciones
Puntas compatibles Puntas de diamante, puntas Amplipur
Espaciado de las puntas ajustable No
Esterilizables en autoclave Esterilizable en autoclave
Color Surtido
Desechables No
Ver más especificaciones
Descripción
Descripción
MyPIPETMAN™ Enterprise pipettes are produced in a series of at least 150 units (in a mix of different volumes). All pipettes inside a series have the same technical features, design, and information printed on the handle.
  • Select your preferred technical features to optimize the use of your pipette and overall productivity in the lab
  • Color choices and printing options on your pipette allow for better organization and identification within your workflow
Especificaciones
Especificaciones
Puntas de diamante, puntas Amplipur
Esterilizable en autoclave
No
Polipropileno, PVDF, PBT
No estéril
No
Surtido
Con textura
MyPipetman™ Enterprise
Pipeta mecánica