Learn More
Sontara™ Multipurpose EC Wipe
Descripción
- All-around industrial wipes family
- Wet, dry or in between, Sontara™ wipes are more versatile than cloth rags, stronger and more durable than paper, and less expensive in use than both
- Made under strict production controls for consistent quality, uniform size and packaging
- Well-suited for the many clean-up and prep tasks in the manufacturing and transportation industry
- Tough and durable for multiple reuses
- In test after test, proven to be among the strongest, wet or dry
- Can be reused over and over, washes easily yet is inexpensive enough to be treated as a disposable
- Absorbent and dry wipe
- Soaks up to several times its weight in water and also absorbs oil and solvents
- Low-linting
Specifikationer
Specifikationer
| Tipo de producto | Wipe |
| Longitud (imperial) | 12 in |
| Longitud (métrico) | 305 mm |
| Anchura (imperial) | 15 in. |
| Anchura (métrico) | 380 mm |
| Material | Wood Pulp, PET |
| Esterilizables en autoclave | No esterilizable en autoclave |
| Color | Turquesa |
| Desechables | Un solo uso |
| Dimensiones (L x An) | 305 x 380 mm (12 x 15 in) |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?