Learn More
MTSS1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA566843
Detalles adicionales : Peso : 0.01000kg
Descripción
Immunogen sequence: TPTVPDSPGYM GPTRAGSEEC VFYTDETASP LAPDLAKASP KRLSLPNTAW GSPSPEAAGY PGAGAEDEQQ QLA Highest antigen sequence identity to the following orthologs - mouse 82%, rat 84%.
Involved in plasma membrane dynamics. Potentiated PDGF-mediated formation of membrane ruffles and lamellipodia in fibroblasts, acting via RAC1 activation (PubMed:14752106). May function in actin bundling (PubMed:14752106). [UniProt]Especificaciones
MTSS1L | |
Polyclonal | |
Unconjugated | |
Mtss1l | |
ABBA; ABBA1; ABBA-1; actin-bundling protein with BAIAP2 homology; actin-bundling with BAIAP2 homology protein 1; LOC92154; metastasis suppressor 1 like; metastasis suppressor 1-like; Mtss1l; MTSS1-like protein; MTSS2; Protein MTSS 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92154 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q765P7 | |
Mtss1l | |
Recombinant Human MTSS1L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only