Learn More
MTG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA558127
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: QHLRKGKATR VGGEPGITRA VMSKIQVSER PLMFLLDTPG VLAPRIESVE TGLKLALCGT VLDHLVGEET MADYLLYTLN KH Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
Mitochondrial GTPase. May be involved in assembly of the large ribosomal subunit.Especificaciones
MTG1 | |
Polyclonal | |
Unconjugated | |
MTG1 | |
Gm169; GTP; GTP binding protein 7 (putative); GTP-binding protein 7; GTP-binding protein 7 (putative); GTPBP7; Mitochondrial GTPase 1; mitochondrial GTPase 1 homolog; mitochondrial GTPase 1 homolog (S. cerevisiae); mitochondrial GTPase 1-like; mitochondrial ribosome associated GTPase 1; mitochondrial ribosome-associated GTPase 1; Mtg1; RP11-108K14.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92170 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BT17 | |
MTG1 | |
Recombinant protein corresponding to Human MTG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only