Learn More
MSRB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA531355
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: U87-MG, MCF-7. Predicted reactivity: Human (99%), Mouse (83%), Rat (85%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MSRB2 catalyzes the reduction of free and protein-bound methionine sulfoxide to methionine.Especificaciones
MSRB2 | |
Polyclonal | |
Unconjugated | |
MSRB2 | |
2310050L06Rik; CBS1; CBS-1; CGI-131; methionine sulfoxide reductase B2; methionine-R-sulfoxide reductase B2, mitochondrial; Mrsb; Msrb; MsrB2; PILB; pilin-like transcription factor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22921 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y3D2 | |
MSRB2 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 132 of Human MSRB2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |