missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS34 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15166-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated75% protein to rRNA composition compared to prokaryotic ribosomes,where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. Alternate splice variants of this gene have been described but their full-length nature has not been determined.Especificaciones
MRPS34 | |
Polyclonal | |
Unconjugated | |
MRPS34 | |
MRP S12, MRP S34, MRPS12, MRPS34, S34mt | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
287126, 65993, 79044 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P82930, Q9JIK9 | |
Mrps34 | |
MRPS34 Fusion Protein Ag7287 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |