missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS23 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18345-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
PHTNS interacts with regulatory subunits that target the enzyme to different cellular locations and change its activity toward specific substrates. Phostensin is a regulatory subunit that targets PP1 to F-actin cytoskeleton.Especificaciones
MRPS23 | |
Polyclonal | |
Unconjugated | |
MRPS23 | |
CGI 138, HSPC329, MRP S23, MRPS23, S23mt | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
360594, 51649, 64656 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VE22, Q9Y3D9 | |
Mrps23 | |
MRPS23 Fusion Protein Ag13144 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |