missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL54 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA538956
Detalles adicionales : Peso : 0.00900kg
Descripción
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. [provided by RefSeq, Jul 2008].Especificaciones
MRPL54 | |
Polyclonal | |
Unconjugated | |
MRPL54 | |
MRPL54 | |
A synthetic peptide derived from the internal region of human MRPL54 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6P161, Q9CPW3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116541, 66047 | |
-20°C | |
Liquid |