Learn More
MRPL50 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA554638
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: MAERSVSGIT RRVFMWTVSG TPCREFWSRF RKEKEPVVVE TVEEKKEPIL VCPPLRSRAY TPPEDLQSRL ESYVKEV Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 67%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a putative 39S subunit protein and belongs to the L47P ribosomal protein family. Pseudogenes corresponding to this gene are found on chromosomes 2p, 2q, 5p, and 10q. [provided by RefSeq, Jul 2008].Especificaciones
MRPL50 | |
Polyclonal | |
Unconjugated | |
MRPL50 | |
39S ribosomal protein L50, mitochondrial; L50mt; mitochondrial 39S ribosomal protein L50; Mitochondrial large ribosomal subunit protein mL50; mitochondrial ribosomal protein L50; MRPL50; MRP-L50 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54534 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N5N7 | |
MRPL50 | |
Recombinant protein corresponding to Human MRPL50 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only