missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL34 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA568064
Detalles adicionales : Peso : 0.01000kg
Descripción
Predicted to react with Mouse samples.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.Especificaciones
MRPL34 | |
Polyclonal | |
Unconjugated | |
Mrpl34 | |
0610007O17Rik; 39S ribosomal protein L34, mitochondrial; D8Bwg1484e; L34mt; Mitochondrial large ribosomal subunit protein bL34m; mitochondrial ribosomal protein L34; MRPL34; MRP-L34; RM34 | |
Rabbit | |
affinity chromatography | |
RUO | |
64981, 94065 | |
-20°C | |
Liquid |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99N91, Q9BQ48 | |
Mrpl34 | |
Synthesized peptide derived from human MRPL34. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |