Learn More
ACROBiosystems Mouse EpCAM / TROP1 Protein, His Tag
Marca: ACROBiosystems EPM-M5227-100UG
Detalles adicionales : Peso : 0.00001kg
Descripción
EpCAM is also known as CO171A, EGP, EGP40,GA7332, KSA, M4S, MIC18, MK1, TROP1, hEGP2, and is a pan-epithelial differentiation antigen that is expressed on almost all carcinomas as 17-1A(mAb) antigen. Its constitutional function is being elucidated. It is intricately linked with the Cadherin-Catenin pathway and hence the fundamental WNT pathway responsible for intracellular signaling and polarity. The epithelial cell adhesion molecule (Ep-CAM) is known to express in most epithelial malignancies and was reported as a tumor marker or a candidate of molecular targeting therapy. Ep-CAM cross signaling with N-cadherin involves Pi3K, resulting in the abrogation of the cadherin adhesion complexes in epithelial cells was reported. And Epithelial cell adhesion molecule (Ep-CAM) recently received increased attention as a prognostic factor in breast cancer.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.5 kDa. The protein migrates as 33-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAH05618.1 | |
28.5 kDa | |
100 µg | |
Gln 24 - Thr 266 | |
EPCAM,TACSTD1,TROP1,CD326,DIAR5,EGP2,EGP314,EGP40,ESA,GA733-2,HNPCC8,HNPCC-8,KS1/4,KSA,M4S1,MIC18,MK1 | |
Unconjugated | |
PBS, pH7.4 | |
HEK293 | |
Mouse |
Powder | |
>98% as determined by SDS-PAGE | |
Mouse EpCAM, His Tag (EPM-M5227) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Thr 266 (Accession # AAH05618.1). | |
-20°C | |
EpCAM | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Bio-Markers and CD Antigens |