missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgA Secondary Antibody, HRP, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marca: Invitrogen SA135467
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody does not cross react with albumin fractions of human serum, human IgG, human IgM or albumin fractions from other species.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Especificaciones
Human IgA | |
Monoclonal | |
1.2 mg/mL | |
PBS with 50% glycerol, 5mg/mL BSA and no preservative; pH 7.5 | |
Purified alpha-chain (Fc) of human IgA | |
1 mL | |
Secondary | |
-20°C | |
IgG1 |
Dot Blot, ELISA, Immunohistochemistry (Frozen) | |
BFG-267 | |
HRP | |
Mouse | |
Ion-exchange chromatography, Size-exclusion - Gel filtration | |
RUO | |
Human | |
Liquid |