Learn More
Novus Biologicals™ Monkey Calcitonin ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Marca:  Novus Biologicals™ NBP3-06761
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18384772
  • 769.96€ / 1 set
Fecha estimada de envío: 25-06-2024
para ver el stock
Especificaciones
15.63 to 1000 pg/mL
ELISA Kit (Colorimetric)
This kit recognizes Monkey CT in samples. No significant cross-reactivity or interference between Monkey CT and analogues was observed.
796
1 Kit
Storage of components varies. See protocol for specific instructions.
Monkey
9.38 pg/mL
Biological Fluids, Cell Culture Supernatant, Plasma, Serum
ELISA, Sandwich ELISA
CALCA
Cancer, Cellular Markers, Neuroscience
ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump