Learn More
Novus Biologicals™ Monkey Calcitonin ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Marca: Novus Biologicals™ NBP3-06761
Detalles adicionales : Peso : 0.00970kg
Especificaciones
15.63 to 1000 pg/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Monkey CT in samples. No significant cross-reactivity or interference between Monkey CT and analogues was observed. | |
796 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Monkey |
9.38 pg/mL | |
Biological Fluids, Cell Culture Supernatant, Plasma, Serum | |
ELISA, Sandwich ELISA | |
CALCA | |
Cancer, Cellular Markers, Neuroscience | |
ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump |