missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17977-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
AQP0 is the most abundant endogenous protein in the plasma membrane of lens fiber cells where it functions not only as a water pore, but it is also involved in fiber-fiber adhesion and is crucial for fiber cell structure and organization. AQP0 contains an additional pore constriction, not seen in any other aquaporin structures, which may be responsible for pore gating. The closed AQP0 pore holds just three water molecules, which are spaced too far apart to form hydrogen bonds with each other. The C-terminal domain of AQP0 undergoes extensive post-translational modification, including many truncations, during lens aging due to the actions of m-calpain, proteases or non-enzymatic mechanisms. These truncation sites may be involved in the development of cataracts.Especificaciones
MIP | |
Polyclonal | |
Unconjugated | |
MIP | |
AQP0, Aquaporin 0, LIM1, MIP, MIP26, MP26 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17339, 25480, 4284 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09011, P30301, P51180 | |
Mip | |
MIP Fusion Protein Ag12369 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |