missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIP-3 alpha Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26527-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Macrophage Inflammatory Protein-3 alpha (MIP-3alpha), also called CCL20, is expressed in the liver, lungs, lymph nodes and peripheral blood lymphocytes. MIP-3alpha is strongly up regulated by inflammatory signals, and down regulated by the anti-inflammatory cytokine IL-10. MIP3alpha signals through the G protein-coupled receptor, CCR6, and acts as a chemoattractant to lymphocytes and dendritic cells.Especificaciones
MIP-3 alpha | |
Polyclonal | |
Unconjugated | |
CCL20 | |
Beta chemokine exodus 1, C C motif chemokine 20, CC chemokine LARC, CCL20, CKb4, LARC, MIP 3 alpha, MIP 3a, MIP3A, MIP-3alpha, SCYA20, Small inducible cytokine A20, ST38 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6364 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P78556 | |
CCL20 | |
MIP-3alpha Fusion Protein Ag24069 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |