missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-4-bromofenilacetato de metilo, 96 %, Thermo Scientific™

Catalog Number 15496527
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1182284-46-3
C11H12BrNO3
286.125
MFCD12923023
LCQMFPAASSGQFD-UHFFFAOYSA-N
methyl 2-2-acetamido-4-bromophenyl acetate, methyl 2-acetamido-4-bromophenylacetate, methyl 2-4-bromo-2-acetamidophenyl acetate, 2-acetamido-4-bromophenylacetic acid methyl ester
46784005
2-(2-acetamido-4-bromofenil)acetato de metilo
CC(=O)NC1=C(C=CC(=C1)Br)CC(=O)OC
Este artículo no se puede devolver. Vea la política de devoluciones
1182284-46-3
C11H12BrNO3
286.125
MFCD12923023
LCQMFPAASSGQFD-UHFFFAOYSA-N
methyl 2-2-acetamido-4-bromophenyl acetate, methyl 2-acetamido-4-bromophenylacetate, methyl 2-4-bromo-2-acetamidophenyl acetate, 2-acetamido-4-bromophenylacetic acid methyl ester
46784005
2-(2-acetamido-4-bromofenil)acetato de metilo
CC(=O)NC1=C(C=CC(=C1)Br)CC(=O)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Methyl 2-acetamido-4-bromophenylacetate
Punto de fusión 126°C to 127°C
Cantidad 250 mg
Formula Weight (peso de la fórmula) 286.13
Porcentaje de pureza 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'