Learn More
2,2-Dimetilacetoacetato de metilo, 99 %, Thermo Scientific Chemicals
CAS | 38923-57-8 |
---|---|
Fórmula molecular | C7H12O3 |
Peso molecular (g/mol) | 144.17 |
Número MDL | MFCD00026232 |
Clave InChI | LVSDLZIEHYYLTC-UHFFFAOYSA-N |
Sinónimo | methyl 2,2-dimethylacetoacetate, methyl2,2-dimethylacetoacetate, butanoic acid, 2,2-dimethyl-3-oxo-, methyl ester, methyl 2,2-dimethyl acetoacetate, 2,2-dimethylacetoacetic acid methyl ester, 2,2-dimethyl-3-oxo-butyric acid methyl ester, 2,2-dimethyl-3-oxobutyric acid, methyl ester, methyl 2,2-dimethyl-3-oxobutyrate, pubchem23356, acmc-20akj7 |
PubChem CID | 96925 |
Nombre IUPAC | 2,2-dimetil-3-oxobutanoato de metilo |
SMILES | COC(=O)C(C)(C)C(C)=O |
Descripción
Pyrimidine was synthesis from methyl 2, 2-dimethylacetoacetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AplicacionesLa pirimidina se sintetizó a partir 2, 2-dimetilacetoacetato de metilo.
Solubilidad
Insoluble en agua.
Notas
Almacenar en un lugar fresco. Mantener el recipiente bien cerrado en un lugar seco y bien ventilado. Almacenar lejos de los agentes oxidantes fuertes.
38923-57-8 | |
144.17 | |
LVSDLZIEHYYLTC-UHFFFAOYSA-N | |
96925 | |
COC(=O)C(C)(C)C(C)=O |
C7H12O3 | |
MFCD00026232 | |
methyl 2,2-dimethylacetoacetate, methyl2,2-dimethylacetoacetate, butanoic acid, 2,2-dimethyl-3-oxo-, methyl ester, methyl 2,2-dimethyl acetoacetate, 2,2-dimethylacetoacetic acid methyl ester, 2,2-dimethyl-3-oxo-butyric acid methyl ester, 2,2-dimethyl-3-oxobutyric acid, methyl ester, methyl 2,2-dimethyl-3-oxobutyrate, pubchem23356, acmc-20akj7 | |
2,2-dimetil-3-oxobutanoato de metilo |
Especificaciones
Methyl 2,2-dimethylacetoacetate | |
0.999 | |
C7H12O3 | |
MFCD00026232 | |
methyl 2,2-dimethylacetoacetate, methyl2,2-dimethylacetoacetate, butanoic acid, 2,2-dimethyl-3-oxo-, methyl ester, methyl 2,2-dimethyl acetoacetate, 2,2-dimethylacetoacetic acid methyl ester, 2,2-dimethyl-3-oxo-butyric acid methyl ester, 2,2-dimethyl-3-oxobutyric acid, methyl ester, methyl 2,2-dimethyl-3-oxobutyrate, pubchem23356, acmc-20akj7 | |
LVSDLZIEHYYLTC-UHFFFAOYSA-N | |
2,2-dimetil-3-oxobutanoato de metilo | |
96925 | |
99% |
38923-57-8 | |
72°C to 74°C (15 mmHg) | |
1.4205 | |
1753940 | |
Insoluble in water. | |
COC(=O)C(C)(C)C(C)=O | |
144.17 | |
144.17 |
Seguridad y manipulación
missing translation for 'einecsNumber' : 254-189-1
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes
RUO – Research Use Only