Learn More
Honeywell Safety Products™ Maxi Welder Cut Gloves
Catalog Number p-7115902
Click to view available options
Tamaño:
8
9
Weld with superior protection using these MIG and arc welding, type B gloves. Honeywell Safety Products™ Maxi Welder Cut Gloves, made of cowhide and Kevlar™, are designed with flexible materials and features for excellent comfort.
  • Cowhide leather/Kevlar™ construction.
  • Winged thumb with yoke for flexibility and comfort.
  • Seamless forefinger.
  • Gauntlet type.
  • Silicone-coated Kevlar™ palm for heat and cut protection.
  • Kevlar™ fleece lining for heat insulation.
  • Inner lining of cotton for comfort and sweat absorption.
  • Water-repellent cowhide grain back.
  • Excellent resistance to contact and radiant heat.
  • Elastic on the back.
  • Kevlar™ stitching.
  • Split leather cuff to protect the forearm from mechanical hazards.
TRUSTED_SUSTAINABILITY
Certificaciones/conformidad CE CATEGORY II Intermediary risks, EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN 407-04 Heat hazards, EN 12477-2000 Welding hazards, EC Category PPE 2, EC Certificate Number 0075/797/162/10/12/0807, EC Attestation Number 0075/797/162/10/12/0807
Clasificación Dexterity: 5
Área revestida Palm
Material del manguito Split Leather
Estilo de corte Gunn
Coating Material Silicone
Cut Resistance Rating 4
Material Grain Leather, Dupont™ Kevlar™ Aramid Fibers
Tipo de producto Welding Gloves
Tamaño 8
Para utilizar con (aplicación) Automotive, Foundry, Iron, Steel, Ship Building, Transportation, Utility and Welding industries.
Estilo de puño de los guantes Gauntlet
Material de revestimiento Cotton
Revestimiento Lined
Cantidad 20/PK
Clasificación de desgarro 2
Longitud (métrico) 38.5 cm