missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mammaglobin A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 23501-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This polyclonal antibody is raised against the C-terminal 71 amino acids of the protein encoded by a cDNA.
Mammaglobin is a gene that is expressed almost exclusively in the normal breast epithelium and human breast cancer. It is a member of the secretoglobin gene family and forms a heterodimer with lipophilin B. It has been suggested that mammaglobin may be a useful marker for breast cancer clinical research. Studies investigating the detection of mRNA by RT PCR from circulating carcinoma cells in the peripheral blood of breast cancer patients have shown that mammaglobin is a highly specific marker and correlates with several prognostic factors, such as lymph node involvement.Especificaciones
Mammaglobin A | |
Polyclonal | |
Unconjugated | |
SCGB2A2 | |
hMAM, Mammaglobin 1, Mammaglobin A, MGB1, SCGB2A2, UGB2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4250 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13296 | |
SCGB2A2 | |
Mammaglobin A Fusion Protein Ag20122 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |