missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysozyme Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2G5A8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66456-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Lysozyme is synthesized predomitly in reactive histiocytes rather than in resting, unstimulated phagocytes.Especificaciones
Lysozyme | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P17897, P61626, Q05820 | |
LYZ, Lyz1, Lyz2 | |
Lysozyme Fusion Protein Ag6917 | |
20 μL | |
Primary | |
Rat, Mouse, Human, Porcine | |
Antibody | |
IgG2a |
Western Blot | |
2G5A8 | |
Unconjugated | |
LYZ | |
1,4 beta N acetylmuramidase C, lysozyme, lysozyme (renal amyloidosis), Lysozyme C, LYZ, LZM | |
Mouse | |
Protein A | |
RUO | |
100157211, 17110, 25211, 4069 | |
-20°C | |
Liquid |