missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G Rat, FITC, Clone: 1A8, enQuireBio™
Rat Monoclonal Antibody
Marca: EnquireBio QAB56-F-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Ly-6G | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Ly6g | |
Protein A/G Purified | |
RUO | |
546644 | |
IgG2a κ |
Flow Cytometry | |
1A8 | |
FITC | |
XP_006521677 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |