missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Luteinizing Hormone / LHCGR Mouse, Clone: LHCGR/1417, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 3973-MSM3-P1
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Luteinizing Hormone / LHCGR | |
Cytoplasmic and Cell Surface | |
LHCGR/1417 | |
Unconjugated | |
NP_000224 | |
Mouse | |
78-85kDa | |
Protein A/G Purified | |
RUO | |
3973 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
LHCGR | |
Recombinant N-terminal fragment of human LHCGR | |
HepG2 or Jurkat cells. Ovarian Carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |