Learn More
LPAM-1 Rat, Biotin, Clone: DATK32, enQuireBio™
Rat Monoclonal Antibody
Marca: EnquireBio QAB73-B-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
LPAM-1 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Protein A/G Purified | |
RUO | |
Store at 2-4°C. |
Flow Cytometry | |
DATK32 | |
Biotin | |
Rat | |
25 μg | |
Primary | |
IgG2a κ |