missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LIN28A Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP1012
Detalles adicionales : Peso : 0.01000kg
Descripción
Lin28 Synthetic Peptide, PEP-1012, from Invitrogen.
This peptide corresponds to 15 amino acids near the carboxy terminus of human Lin28.PEP-1012 can be used as a blocking peptide with polyclonal antibody PA5-20898.Especificaciones
79727 | |
purified | |
Human LIN28A | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9H9Z2 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
LIN28A | |
0.02% sodium azide | |
50 μg | |
PBS |