missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LASP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10515-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of a LIM protein subfamily characterized by a LIM motif and a domain of Src homology region 3. The encoded protein functions as an actin-binding protein and possibly in cytoskeletal organization.Especificaciones
LASP1 | |
Polyclonal | |
Unconjugated | |
LASP1 | |
Lasp 1, LASP1, LIM and SH3 domain protein 1, LIM and SH3 protein 1, MLN 50, MLN50 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16796, 29278, 3927 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14847, Q61792, Q99MZ8 | |
LASP1 | |
LASP1 Fusion Protein Ag0800 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |