Learn More
Camlab™ Label Laser
Marca: Camlab™ RTP100
Detalles adicionales : Peso : 0.36360kg
Descripción
Self adhesive, smear resistant with a high degree of chemical resistance- Available as Hi-Low strips and Hi-Low labels
- Both types suitable for use between -85°C to 100°C
- Designed for use with thermal transfer and laser printers
- Hand feeding recommended
Especificaciones
Label Laser | |
33 mm | |
13 mm | |
1980/Pk. |