missing translation for 'onlineSavingsMsg'
Learn More

Transforming Growth Factor α (TGFα) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ Transforming Growth Factor α (TGFα) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Marca:  Epredia MS-670-PABX

Detalles adicionales : Peso : 0.00180kg

 Ver más versiones de este producto

Código de producto. 12612007

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

TGFα (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.

Host Species: Mouse

Clone: MF9

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Rat and Mouse. Others not known.

Epitope: C-terminal

Immunogen: A 17-amino acid synthetic peptide from the carboxyl-terminus of rat TGFα

Molecular Weight: 6kDa

Positive Control: Recombinant TGFα protein, A2058 cells

Cellular Localization: Cytoplasmic Ab-1 shows no cross-reaction with EGF and the neuropeptide synenkephalin. Ab-1 is excellent for staining of formalin-fixed, paraffin-embedded tissues. The staining with Ab-1 is completely blocked by the peptide used for raising this antibody.

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Transforming Growth Factor alpha (TGFalpha) Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Signal Transduction
Bovine, Human, Mouse, Rat
IgG2a κ
Western Blot
MF9
Unconjugated
A 17-amino acid synthetic peptide from the carboxyl-terminus of Rat TGFalpha
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales