missing translation for 'onlineSavingsMsg'
Learn More

TIMP-2 Ab-5 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ TIMP-2 Ab-5, Mouse Monoclonal Antibody.

Marca:  Epredia MS-1485-PABX

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12699906

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.

Host Species: Mouse

Clone: 3A4

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse, Rat, Rabbit, Cow and Guinea Ppig. Others not known.

Epitope: N-terminal

Immunogen: A synthetic peptide from the N-terminal region of human TIMP-2 protein

Molecular Weight: 21kDa

Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells; Pancreatic or colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting
  • Immunoprecipitation (Not suitable)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

TIMP-2 Ab-5
Monoclonal
1mg/mL
Mouse
200 μL
Proteases And Inhibitors
Bovine, Guinea Pig, Human, Mouse, Rabbit, Rat
IgG2a κ
Immunohistochemistry (Paraffin), Western Blot
3A4
Unconjugated
A synthetic peptide from the N-terminal region of human TIMP-2 protein
RUO
Primary
Concentrated
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali