Learn More
TIMP-2 Ab-5 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ TIMP-2 Ab-5, Mouse Monoclonal Antibody.
Marca: Epredia MS-1485-P1
Detalles adicionales : Peso : 1.00000kg
Descripción
TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.Host Species: Mouse
Clone: 3A4
Isotype: IgG2a/κ
Species Reactivity: Human, Mouse, Rat, Rabbit, Cow and Guinea Ppig. Others not known.
Epitope: N-terminal
Immunogen: A synthetic peptide from the N-terminal region of human TIMP-2 protein
Molecular Weight: 21kDa
Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells; Pancreatic or colon CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
- Western Blotting
- Immunoprecipitation (Not suitable)
Note:
USA: RUO; Int'l: RUO
Especificaciones
TIMP-2 Ab-5 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Proteases And Inhibitors | |
Bovine, Guinea Pig, Human, Mouse, Rabbit, Rat | |
IgG2a κ |
Immunohistochemistry (Paraffin), Western Blot | |
3A4 | |
Unconjugated | |
A synthetic peptide from the N-terminal region of human TIMP-2 protein | |
RUO | |
Primary | |
Concentrated |