Learn More
Epredia™ Lab Vision™ Synaptophysin (Neuroendocrine Marker) Rabbit Monoclonal Antibody
Synaptophysin (Neuroendocrine Marker), Rabbit MAb is part of a comprehensive ab portfolio. Studies show rabbit mabs are more sensitive and specific than other available markers.
Marca: Epredia™ RM-9111-R7
Detalles adicionales : Peso : 0.01200kg
Descripción
This antibody recognizes a protein of 38kDa, identified as synaptophysin. It labels normal neuroendocrine cells of human adrenal medulla, carotid body, skin, pituitary gland, thyroid, lung, pancreas, gastrointestinal mucosa, Paneth's cells in the gastrointestinal tract and of gastric parietal cells. Neurons in the brain, spinal cord, and retina are also labeled. In combination with anti-chromogranin A and anti-NSE, Ab to synaptophysin is very useful in the identification of normal neuroendocrine cells and neuroendocrine neoplasms.Host Species: Rabbit
Clone: SP11
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: C-terminus
Immunogen: A synthetic peptide of human synaptophysin
Molecular Weight: 38kDa
Positive Control: IMR5 cells, Pancreas or Pheochromocytoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Especificaciones
Synaptophysin (Neuroendocrine Marker) | |
Monoclonal | |
Unconjugated | |
A synthetic peptide of human synaptophysin | |
IVD | |
Primary | |
Prediluted |
Western Blot | |
SP11 | |
Rabbit | |
7 mL | |
Neuroscience | |
Human | |
IgG |